missing translation for 'onlineSavingsMsg'
Learn More

Cyproterone acetate, 98%, Thermo Scientific Chemicals

Product Code. 15549483 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15549483

Brand: Thermo Scientific Acros 459542500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 427-51-0
Molecular Formula C24H29ClO4
Molecular Weight (g/mol) 416.94
InChI Key UWFYSQMTEOIJJG-FDTZYFLXSA-N
Synonym cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385
PubChem CID 9880
ChEBI CHEBI:50743
SMILES CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C

Specifications

Melting Point 207.0°C to 215.0°C
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (80°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, methanol, slightly soluble in ethanol
Formula Weight 416.94
Percent Purity 98%
Chemical Name or Material Cyproterone acetate
compliance-icons compliance-icons
Product Identifier
  • Cyproterone acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.