missing translation for 'onlineSavingsMsg'
Learn More

Cyproheptadine N-Oxide, TRC

Product Code. 30137225
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137225 1 g 1g
30011601 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137225 Supplier Toronto Research Chemicals Supplier No. TRCC9895501G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100295-63-4
Molecular Formula C21 H21 N O
Molecular Weight (g/mol) 303.4
Synonym 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-piperidine 1-Oxide, Cyprohepatadine N-Oxide
IUPAC Name 4-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-1-methyl-1-oxidopiperidin-1-ium
SMILES C[N+]1([O-])CCC(=C2c3ccccc3C=Cc4ccccc24)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H21NO/c1-22(23)14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21/h2-11H,12-15H2,1H3
Formula Weight 303.1623
Chemical Name or Material Cyproheptadine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.