missing translation for 'onlineSavingsMsg'
Learn More

Cyproheptadine Hydrochloride Sesquihydrate, TRC

Product Code. 30179793
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179793 100 mg 100mg
30180266 1 g 1g
30180282 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179793 Supplier Toronto Research Chemicals Supplier No. TRCC989200100MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41354-29-4
Molecular Formula 2 C21 H21 N . 2 Cl H . 3 H2 O
Molecular Weight (g/mol) 701.76
Synonym Cyproheptadine Hydrochloride Sesquihydrate, Piperidine, 4-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-, hydrochloride, hydrate (2:3) (9CI)
IUPAC Name 4-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-1-methylpiperidine;trihydrate;dihydrochloride
SMILES O.O.O.Cl.Cl.CN1CCC(=C2c3ccccc3C=Cc4ccccc24)CC1.CN5CCC(=C6c7ccccc7C=Cc8ccccc68)CC5

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C21H21N.2ClH.3H2O/c2*1-22-14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21;;;;;/h2*2-11H,12-15H2,1H3;2*1H;3*1H2
Formula Weight 700.3198
Chemical Name or Material Cyproheptadine Hydrochloride Sesquihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.