missing translation for 'onlineSavingsMsg'
Learn More

Cyproheptadine hydrochloride sesquihydrate, MedChemExpress

Product Code. 16418656
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418656 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418656 Supplier Medchem Express Supplier No. HYB1165

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Cyproheptadine hydrochloride sesquihydrate is an antihistamine and is an antagonist of serotonin and histamine2.

Cyproheptadine hydrochloride sesquihydrate is an antihistamine and is an antagonist of serotonin and histamine2.

Specifications

Chemical Name or Material Cyproheptadine hydrochloride sesquihydrate
CAS 41354-29-4
Color White
Purity Grade Notes Research
Molecular Formula C21H21N.3/2H2O.HCl
Quantity 100 mg
Solubility Information DMSO : 33.33 mg/mL (94.99 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.91 mM; Need ultrasonic)
SMILES CN1CC/C(CC1)=C2C3=CC=CC=C3C=CC4=CC=CC=C/24.[H]Cl.[1.5H2O]
Molecular Weight (g/mol) 350.88
Formula Weight 350.88
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.