missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyproheptadine hydrochloride sesquihydrate

Product Code. 15891042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15891042 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15891042 Supplier Thermo Scientific Chemicals Supplier No. 462670050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41354-29-4 | C42H50Cl2N2O3 | 701.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 41354-29-4
Melting Point 165.0°C
Color Clear Colorless to Light Yellow
Molecular Formula C42H50Cl2N2O3
MDL Number MFCD27967225,MFCD00012538,MFCD00012538,MFCD27967225,MFCD00242817
Quantity 5 g
Solubility Information Solubility aq. soln.: very slightly soluble. Other solubilities: very soluble in methanol, soluble in chloroform, slightly soluble in ethanol
InChI Key ZEAUHIZSRUAMQG-UHFFFAOYSA-N
SMILES [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12
IUPAC Name dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
Molecular Weight (g/mol) 701.77
Formula Weight 350.88
Chemical Name or Material Cyproheptadine hydrochloride sesquihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.