missing translation for 'onlineSavingsMsg'
Learn More

Cyproconazole

Product Code. 15455409
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455409 10 g 10g
15445409 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455409 Supplier Thermo Scientific Alfa Aesar Supplier No. J63142.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Triazole fungicide which causes hepatocellular adenomas and carcinomas | CAS: 94361-06-5 | C15H18ClN3O | 291.779 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Cyproconazole is used to inhibit demeythylation in fungal sterol synthesis. And it is used on greenhouse and field-grown roses and as a wood preservative, surface application or pressure treatment of wood for above-ground uses, including siding, plywood, millwork, shingles, lumber.

Solubility
Soluble in acetone, ethanol, xylene, and dimethyl sulfoxide.

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94361-06-5
Molecular Formula C15H18ClN3O
Molecular Weight (g/mol) 291.779
MDL Number MFCD01678672
InChI Key UFNOUKDBUJZYDE-UHFFFAOYSA-N
PubChem CID 86132
ChEBI CHEBI:83748
IUPAC Name 2-(4-chlorophenyl)-3-cyclopropyl-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(C1CC1)C(CN2C=NC=N2)(C3=CC=C(C=C3)Cl)O

Specifications

Type Cyproconazole
Physical Form Crystalline
Melting Point 108°C
Color Brown
Quantity 10 g
Flash Point >100°C (212°F)
UN Number UN3077
Beilstein 8396421
Boiling Point >250°C
Merck Index 14,2771
Solubility Information Soluble in acetone,ethanol,xylene,and dimethyl sulfoxide.
Formula Weight 291.78
Chemical Name or Material Cyproconazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyproconazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Cyproconazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.