missing translation for 'onlineSavingsMsg'
Learn More

Cyproconazole, MedChemExpress

Product Code. 16400758
Encompass_Preferred
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 16400758

Brand: Medchem Express HYA0277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyproconazole is a triazole fungicide that is used agriculturally for protection of crops against a wide variety of fungal pathogens.

Cyproconazole is a triazole fungicide that is used agriculturally for protection of crops against a wide variety of fungal pathogens.

Specifications

Chemical Name or Material Cyproconazole
Physical Form Solid
CAS 94361-06-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C15H18ClN3O
Solubility Information DMSO : 100 mg/mL (342.72 mM; Need ultrasonic)
SMILES ClC1=CC=C(C(C(C)C2CC2)(O)CN3N=CN=C3)C=C1
Molecular Weight (g/mol) 291.78
Formula Weight 291.78
Percent Purity 98.62%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.