missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP8B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102021
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cytochrome P450 8B1.
CYP8B1 (sterol 12-alpha-hydroxylase) is a member of the cytochrome P450 superfamily of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds. CYP8B1 is highly expressed in liver and is an important enzyme for bile acid synthesis. Specifically, CYP8B1 moderates the ratio of cholic acid over chenodeoxycholic acid to control the solubility of cholesterol. The gene encoding human CYP8B1 maps to chromosome 3p22.1. The CYP8B1 gene encodes a 501-amino acid protein and does not contain any introns. The CYP8B1 gene promoter is transactivated by hepatocyte nuclear factor 4a. In mice, disruption of the CYP8B1 gene prevents the synthesis of cholate, which is a primary bile acid.
Specifications
| CYP8B1 | |
| Polyclonal | |
| Unconjugated | |
| CYP8B1 | |
| 7 alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase; 7-alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase; 7-alpha-hydroxycholest-4-en-3-one 12-alpha-hydroxylase; CP8B; CYP12; Cyp8b1; CYPVIIIB1; cytochrome P450 8B1; cytochrome P450 8b1 sterol 12 alpha-hydrolase; cytochrome P450 family 8 subfamily B member 1; cytochrome P450, 8b1, sterol 12 alpha-hydrolase; cytochrome P450, family 8, subfamily B, polypeptide 1; cytochrome P450, subfamily VIIIB (sterol 12-alpha-hydroxylase), polypeptide 1; sterol 12-alpha-hydrolase; sterol 12-alpha-hydroxylase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13124, 1582 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88962, Q9UNU6 | |
| CYP8B1 | |
| A synthesized peptide derived from human CYP8B1(Accession Q9UNU6), corresponding to amino acid residues G371-K421. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction