missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP8B1 Antibody (M15-P3B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £302.00
Specifications
| Antigen | CYP8B1 |
|---|---|
| Clone | M15-P3B7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679656
|
Novus Biologicals
NBP2-50218-0.025ml |
0.025 mL |
£159.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621538
|
Novus Biologicals
NBP2-50218 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP8B1 Monoclonal antibody specifically detects CYP8B1 in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP8B1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| PBS | |
| 1582 | |
| IgG1 κ | |
| Protein A purified |
| M15-P3B7 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CP8B, CYP127 alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, CYPVIIIB1, Cytochrome P450 8B1, cytochrome P450, family 8, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIIB (sterol 12-alpha-hydroxylase), polypeptide 1,7-alpha-hydroxycholest-4-en-3-one 12-alpha-hydroxylase, EC 1.14.13.95, FLJ17826,7-alpha-hydroxy-4-cholesten-3-one 12-alpha-hydroxylase, Sterol 12-alpha-hydroxylase | |
| Peptide sequence- QPSHDVRFR (amino acids 486 - 494) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title