missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP7B1, Mouse anti-Human, Clone: 2B11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009420-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: CDNNKFISELRDDFLKFDDKFAYLVSNIPIELLGNVKSIREKIIKCFSSEKLAKMQGWSEVFQSRQDVLEKYYVHEDLEIGAHHSpecifications
| CYP7B1 | |
| Monoclonal | |
| Unconjugated | |
| cytochrome P450, family 7, subfamily B, polypeptide 1 | |
| CBAS3/CP7B/SPG5A | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2B11 | |
| In 1x PBS, pH 7.4 | |
| NM_004820 | |
| CYP7B1 | |
| CYP7B1 (NP_004811, 203 a.a. ∼ 286 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9420 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction