missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP7B1 Monoclonal Antibody (OTI7H10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524898
This item is not returnable.
View return policy
Description
CYP7B1 Monoclonal Antibody for Western Blot
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway of extrahepatic tissues, which converts cholesterol to bile acids. This enzyme likely plays a minor role in total bile acid synthesis, but may also be involved in the development of atherosclerosis, neurosteroid metabolism and sex hormone synthesis.
Specifications
| CYP7B1 | |
| Monoclonal | |
| 0.63 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75881 | |
| Cyp7b1 | |
| Recombinant protein expressed in E.coli corresponding to amino acids 350-506 of Cyp7b1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI7H10 | |
| Unconjugated | |
| Cyp7b1 | |
| 24-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 25-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; AW261589; CBAS3; CP7B; CYP7B1; Cytochrome P450 7B1; cytochrome P450 family 7 subfamily B member 1; cytochrome P450, 7b1; cytochrome P450, family 7, subfamily b, polypeptide 1; cytochrome P450, subfamily 7B, polypeptide 1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; D3Ertd552e; hct-1; hippocampal transcript 1 protein; Oxysterol 7-alpha-hydroxylase; SPG5A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9420 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction