missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP7B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35797-100ul
This item is not returnable.
View return policy
Description
CYP7B1 Polyclonal antibody specifically detects CYP7B1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CYP7B1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| CBAS3, CP7B, Cytochrome P450 7B1, cytochrome P450, family 7, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1,25-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.100, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase, spastic paraplegia 5A (autosomal recessive), SPG5A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CYP7B1 (NP_004811.1).,, Sequence:, VIVCDNNKFISELRDDFLKFDDKFAYLVSNIPIELLGNVKSIREKIIKCFSSEKLAKMQGWSEVFQSRQDVLEKYYVHEDLEIGAHHLGFLWASVANTIPT | |
| 100 μL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
| 9420 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction