missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP7A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10795-100UL
This item is not returnable.
View return policy
Description
CYP7A1 Polyclonal specifically detects CYP7A1 in Human samples. It is validated for Western Blot.
Specifications
| CYP7A1 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| Cholesterol 7-alpha-hydroxylase, cholesterol 7-alpha-monooxygenase, CP7A, CYP7MGC126826, CYPVII, Cytochrome P450 7A1, cytochrome P450, family 7, subfamily A, polypeptide 1, cytochrome P450, subfamily VIIA (cholesterol 7 alpha-monooxygenase), cytochrome P450, subfamily VIIA polypeptide 1, EC 1.14.13, EC 1.14.13.17, MGC138389, polypeptide 1 | |
| The immunogen is a synthetic peptide directed towards the middle region of human CYP7A1 (NP_000771.2). Peptide sequence DPEIYPDPLTFKYDRYLDENGKTKTTFYCNGLKLKYYYMPFGSGATICPG | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1581 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction