missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP51A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38236-100ul
This item is not returnable.
View return policy
Description
CYP51A1 Polyclonal antibody specifically detects CYP51A1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CYP51A1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| CP51, CYP51Cytochrome P450LI, CYPL1, CYPLI, Cytochrome P450 51A1, cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, EC 1.14.13.70, lanosterol 14-alpha demethylase, lanosterol 14-alpha-demethylase, LDMCytochrome P45014DM, P450-14DM, P450L1,51 (lanosterol 14-alpha-demethylase), Sterol 14-alpha demethylase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-509 of CYP51A1 (NP_000777.1).,, Sequence:, LTASHCLHGKEIRSQLNEKVAQLYADLDGGFSHAAWLLPGWLPLPSFRRRDRAHREIKDIFYKAIQKRRQSQEKIDDILQTLLDATYKDGRPLTDDEVAGMLIGLLLAGQHTSSTTSAWMGFFLARDKTLQKKCYLEQKTVCGENLPPLTYDQLKDLNLLDRCIKETLRLRPPIMIMMRMARTPQTVAGYTIPPGHQVCVSPTVNQRLKDSWVERLDFNPDRYLQDNPASGEKFAYVPFGAGRHRCIGENFAYVQIKTIWSTMLRLYEFDLIDGYFPTVNYTTMIHTPENPVIRYKRRSK | |
| 100 μL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1595 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction