missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP51A1 Antibody (N6-P2H5*G8), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50216DL594
This item is not returnable.
View return policy
Description
CYP51A1 Monoclonal antibody specifically detects CYP51A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| CYP51A1 | |
| Monoclonal | |
| DyLight 594 | |
| CP51, CYP51Cytochrome P450LI, CYPL1, CYPLI, Cytochrome P450 51A1, cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, EC 1.14.13.70, lanosterol 14-alpha demethylase, lanosterol 14-alpha-demethylase, LDMCytochrome P45014DM, P450-14DM, P450L1,51 (lanosterol 14-alpha-demethylase), Sterol 14-alpha demethylase | |
| Ovalbumin-conjugated synthetic peptide NPVIRYKRRS (C-terminal sequence). | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1595 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| N6-P2H5*G8 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction