missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4Z1 Antibody (N7-P2G5*D8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50215H
This item is not returnable.
View return policy
Description
CYP4Z1 Monoclonal specifically detects CYP4Z1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| CYP4Z1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CYP4Z1 | |
| Ovalbumin-conjugated synthetic peptide KLAPDHSRPP (C-terminal sequence) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, metabolism | |
| 199974 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| N7-P2G5*D8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CYP4A20, CYPIVZ1, cytochrome P450 4Z1, cytochrome P450, family 4, subfamily Z, polypeptide 1, EC 1.14.14.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction