missing translation for 'onlineSavingsMsg'
Learn More

CYP4Z1 Antibody (N7-P2G5*D8), CoraFluor™ 1, Novus Biologicals™

Product Code. 30149470
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30149470 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30149470 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP250215CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CYP4Z1 Monoclonal antibody specifically detects CYP4Z1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP4Z1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone N7-P2G5*D8
Conjugate CoraFluor 1
Formulation PBS
Gene Alias CYP4A20, CYPIVZ1, cytochrome P450 4Z1, cytochrome P450, family 4, subfamily Z, polypeptide 1, EC 1.14.14.1
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KLAPDHSRPP (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 199974
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.