missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4V2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556061
This item is not returnable.
View return policy
Description
Immunogen sequence: AQSNDDSEYV RAVYRMSEMI FRRIKMPWLW LDLWYLMFKE GWEHKKSLQI LHTFTNSVIA ERANEMNANE DCRGDGRGSA PSKNKRRAFL DLLLSVTDDE GNRLSHEDIR EEVDTFMFEG HDTTAAAINW SLYLLGSNPE VQK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
This gene encodes a member of the cytochrome P450 hemethiolate protein superfamily which are involved in oxidizing various substrates in the metabolic pathway. It is implicated in the metabolism of fatty acid precursors into n-3 polyunsaturated fatty acids. Mutations in this gene result in Bietti crystalline corneoretinal dystrophy. [provided by RefSeq, Jul 2008].Specifications
CYP4V2 | |
Polyclonal | |
Unconjugated | |
CYP4V2 | |
Cyp4v3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZWL3 | |
CYP4V2 | |
Recombit protein corresponding to Human CYP4V2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only