missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CYP4F3 Monoclonal antibody specifically detects CYP4F3 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | CYP4F3 |
| Applications | ELISA, Western Blot, Sandwich ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4E11 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_000887 |
| Gene Alias | CYP4F, CYPIVF3, cytochrome P-450, Cytochrome P450 4F3, cytochrome P450, family 4, subfamily F, polypeptide 3, cytochrome P450, subfamily IVF, polypeptide 3 (leukotriene B4 omegahydroxylase), Cytochrome P450-LTB-omega, EC 1.14.13.30, leukotriene B4 omega hydroxylase, Leukotriene-B(4) 20-monooxygenase 2, leukotriene-B(4) omega-hydroxylase 2, leukotriene-B4 20-monooxygenase, LTB4HCPF3 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?