missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP4F22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103399
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cytochrome P450 4F22.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This gene is part of a cluster of cytochrome P450 genes on chromosome 19 and encodes an enzyme thought to play a role in the 12(R)-lipoxygenase pathway. Mutations in this gene are the cause of ichthyosis lamellar type 3.
Specifications
| CYP4F22 | |
| Polyclonal | |
| Unconjugated | |
| CYP4F22 | |
| ARCI5; CYP4F22; Cytochrome P450 4F22; cytochrome P450 family 4 subfamily F member 22; cytochrome P450, family 2, subfamily E, polypeptide 2 homolog; cytochrome P450, family 4, subfamily F, polypeptide 22; INLNE; LI3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 126410 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6NT55 | |
| CYP4F22 | |
| A synthesized peptide derived from human CYP4F22(Accession Q6NT55), corresponding to amino acid residues S479-P529. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction