missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP4B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569151
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Goat: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined. Two transcript variants encoding slightly different isoforms have been found for this gene.
Specifications
| CYP4B1 | |
| Polyclonal | |
| Unconjugated | |
| CYP4B1 | |
| CYP4B1; Cyp4b-1; CYP4B1 isoform; CYP4B1-like isozyme short form; CYPIVB1; cytochrome P450 4B1; cytochrome P450 family 4 subfamily B member 1; cytochrome P450 isozyme 5; cytochrome P-450 isozyme 5; Cytochrome P450 L-2; cytochrome P450, family 4, subfamily B, polypeptide 1; cytochrome P450, subfamily 4B, polypeptide 1; cytochrome P450, subfamily IV B, polypeptide 1; cytochrome P450, subfamily IVB, polypeptide 1; cytochrome P450-HP; microsomal monooxygenase; P450 L-2; P-450HP; P450-isozyme 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13120 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q64462 | |
| CYP4B1 | |
| synthetic peptide directed towards the C-terminal region of mouse CYP4B1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction