missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP3A7 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £380.00
Specifications
| Antigen | CYP3A7 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17960453
![]() |
Invitrogen™
PA5145345 |
200 μL |
£380.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17990843
|
Invitrogen™
PA5145346 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene, CYP3A7, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme hydroxylates testosterone and dehydroepiandrosterone 3-sulphate, which is involved in the formation of estriol during pregnancy. The enzyme also metabolizes some drugs such as aflatoxin B1. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Transcript variants have been described, but it is not known whether these transcripts are normally produced.Specifications
| CYP3A7 | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P24462 | |
| 1551 | |
| Recombinant Human CYP3A7 protein, Pro344-Asp497 (Accession #P24462). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| aryl hydrocarbon hydroxylase; CP37; CYP3A7; CYPIIIA7; Cytochrome P450 3A7; cytochrome P450 family 3 subfamily A member 7; cytochrome P450, family 3, subfamily A, polypeptide 7; cytochrome P450, subfamily IIIA, polypeptide 7; cytochrome P450-HFLA; flavoprotein-linked monooxygenase; microsomal monooxygenase; P-450(HFL33); P-450111A7; P450-HFLA; P450HLp2; xenobiotic monooxygenase | |
| CYP3A7 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| CYP3A7 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title