missing translation for 'onlineSavingsMsg'
Learn More

CYP3A7 Mouse anti-Human, Clone: F19 P2 H2, Novus Biologicals™

Product Code. 18346402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346402 200 μg 200µg
18302353 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18346402 Supplier Bio-Techne Supplier No. NBP311914200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CYP3A7 Monoclonal antibody specifically detects CYP3A7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP3A7
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone F19 P2 H2
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry
Formulation PBS
Gene Alias CYPIIIA7, cytochrome P450, family 3, subfamily A, polypeptide 7, Cytochrome P450-HFLA, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, subfamily IIIA, polypeptide 7, xenobiotic monooxygenase
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human CYP3A7 protein and mouse myeloma Ag8563 cells. (Uniprot: Q9H241)
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1551
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.