missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP3A4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142217
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Additional Information: This antibody is expected to recognise both reported isoforms (NP_059488.2; NP_001189784.1) and it is not expected to cross-react with 3A5, 3A7 ot 3A43. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs in use today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4. Alternatively spliced transcript variants encoding different isoforms have been identified.
Tekniske data
| CYP3A4 | |
| Polyclonal | |
| Unconjugated | |
| CYP3A4 | |
| 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; cholesterol 25-hydroxylase; CP33; CP34; CYP; CYP3A; CYP3A3; CYP3A4; CYPIIIA3; CYPIIIA4; Cytochrome P450 3A3; cytochrome P450 3A4; cytochrome P450 family 3 subfamily A member 4; cytochrome P450 HLp; Cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450-PCN1; glucocorticoid-inducible P450; HLP; MGC126680; NF-25; Nifedipine oxidase; P450; P450C3; P450-III, steroid inducible; P450PCN1; P450-PCN1; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1576 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P08684 | |
| CYP3A4 | |
| Peptide with sequence C-KESRLEDTQKHR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion