missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP3A4/CYP3A5 Monoclonal Antibody (F24 P2 B10)
GREENER_CHOICE

Product Code. 11595742
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11595742 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11595742 Supplier Invitrogen™ Supplier No. MA3032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA3-032 detects Cytochrome P450 3A4/3A5 from human cell line HepG2, and from bovine liver and pancreas samples. MA3-032 has been successfully used in Western blot applications. By Western blot, this antibody detects a ~54 kDa band representing Cytochrome P450 3A4/3A5. The MA3-032 immunogen is a synthetic peptide corresponding to residues E S R D G T V S G A, conjugated to ovalbumin.

CYP3A5 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. The enzyme metabolizes drugs such as nifedipine and cyclosporine as well as the steroid hormones testosterone, progesterone and androstenedione.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP3A4/CYP3A5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F24 P2 B10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene CYP3A4
Gene Accession No. P08684, P20815
Gene Alias 1,8-cineole 2-exo-monooxygenase; albendazole monooxygenase; Albendazole monooxygenase (sulfoxide-forming); Albendazole sulfoxidase; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP33; CP34; CP35; CYP3A; CYP3A28; CYP3A3; CYP3A4; CYP3A5; CYPIIIA28; CYPIIIA3; CYPIIIA4; CYPIIIA5; cytochrome P450; cytochrome P450 3A28; cytochrome P450 3A3; Cytochrome P450 3A4; cytochrome P450 3A5; cytochrome P450 family 3 subfamily A member 4; cytochrome P450 family 3 subfamily A member 5; cytochrome P450 HLp; cytochrome P450 HLp2; cytochrome P450 NF-25; cytochrome P450, family 3, subfamily A, polypeptide 4; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; cytochrome P450, subfamily IIIA, polypeptide 4; cytochrome P450-PCN1; Cytochrome P450-PCN3; flavoprotein-linked monooxygenase; glucocorticoid-inducible P450; HLP; HLp2; microsomal monooxygenase; NF-25; Nifedipine oxidase; P450C3; P450-III, steroid inducible; P450PCN1; P450PCN3; P450-PCN3; PCN3; Quinine 3-monooxygenase; taurochenodeoxycholate 6-alpha-hydroxylase; Unknown (protein for MGC:127840); xenobiotic monooxygenase
Gene Symbols CYP3A4, CYP3A5
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues E S R D G T V S G A, conjugated to ovalbumin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1576, 1577, 282214, 507988, 526682
Target Species Bovine, Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.