missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2U1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566722
This item is not returnable.
View return policy
Description
Immunogen sequence: EATIMEVQRLT VVVPLAIPHM TSENTVLQGY TIPKGTLILP NLWSVHRDPA IWEKPEDFYP NRFLDD Highest antigen sequence identity to the following orthologs - mouse 85%, rat 84%.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a hydroxylase that metabolizes arachidonic acid, docosahexaenoic acid, and other long chain fatty acids.
Specifications
| CYP2U1 | |
| Polyclonal | |
| Unconjugated | |
| CYP2U1 | |
| 8430436A10Rik; Cyp2u1; Cytochrome P450 2U1; cytochrome P450 family 2 subfamily U member 1; cytochrome P450, 2u1; cytochrome P450, family 2, subfamily U, polypeptide 1; P450TEC; spastic paraplegia 49; SPG49; SPG56 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z449 | |
| CYP2U1 | |
| Recombinant Human CYP2U1. Recombinant protein control fragment (Product #RP-107375). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction