missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2U1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579133
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA Cell, MCF-7 Cell, MM453 Cell, COLO320 Cell, HT1080 Cell. IHC: Human Intestinal Cancer Tissue. ICC/IF: HELA Cell.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a hydroxylase that metabolizes arachidonic acid, docosahexaenoic acid, and other long chain fatty acids.
Specifications
| CYP2U1 | |
| Polyclonal | |
| Unconjugated | |
| CYP2U1 | |
| 8430436A10Rik; Cyp2u1; Cytochrome P450 2U1; cytochrome P450 family 2 subfamily U member 1; cytochrome P450, 2u1; cytochrome P450, family 2, subfamily U, polypeptide 1; P450TEC; spastic paraplegia 49; SPG49; SPG56 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113612 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q7Z449 | |
| CYP2U1 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human CYP2U1 (457-473aa EKPEDFYPNRFLDDQGQ). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction