missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2R1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529081
This item is not returnable.
View return policy
Description
Predicted reactivity: Mouse (90%), Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor. A mutation in CYP2R1 gene has been associated with selective 25-hydroxyvitamin D deficiency.
Specifications
| CYP2R1 | |
| Polyclonal | |
| Unconjugated | |
| Cyp2r1 | |
| Cyp2r1; Cytochrome P450 2R1; cytochrome P450 family 2 subfamily R member 1; cytochrome P450, 2r1; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; vitamin D 25-hydroxylase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 120227, 244209, 361631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q6VVW9, Q6VVX0 | |
| Cyp2r1 | |
| Recombinant protein encompassing a sequence within the center region of human CYP2R1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction