Learn More
Invitrogen™ CYP2R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587927
Description
Immunogen sequence: VVYDFLSRLI EKASVNRKPQ LPQHFVDAYL DEMDQGKNDP SSTFSKENLI FSVGELIIAG TETTTNVLRW AILFMALYPN IQGQVQKEID LIMGPNGKPS WDDKCKMPYT EAVLHEVLRF CNIVPLGIFH ATSEDAVVRG YSIPKGTTVI TNLYSVHFDE KYWRDPEVFH PERFLDSSGY FAKKEALVPF SLGRRHCLGE HLARMEMFLF FTALLQRFHL HFPHELVPDL KPRLGMTLQP QPYLICAERR; Positive Samples: Mouse testis, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Microsome membrane, Peripheral membrane protein.
CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor. A mutation in CYP2R1 gene has been associated with selective 25-hydroxyvitamin D deficiency.
Specifications
| CYP2R1 | |
| Polyclonal | |
| Unconjugated | |
| Cyp2r1 | |
| Cyp2r1; Cytochrome P450 2R1; cytochrome P450 family 2 subfamily R member 1; cytochrome P450, 2r1; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; vitamin D 25-hydroxylase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 244209, 361631 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.58 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6VVW9 | |
| Cyp2r1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 252-501 of human CYP2R1 (NP_0787902). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.