missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CYP2J2 BACULOSOMES™ Plus Reagent, rHuman

Product Code. 15471204
Change view
Click to view available options
Quantity:
0.5 nmol
Unit Size:
0.5nmol
Product Code. Quantity unitSize
15471204 0.5 nmol 0.5nmol
1 options
This item is not returnable. View return policy

Product Code. 15471204

Brand: Thermo Scientific™ PV6139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase.

Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase. For this particular isozyme (CYP2J2), human cytochrome b5 is also included.

  • Single human P450 isozyme for detailed drug metabolism studies
  • High specific activity increases dynamic range and sensitivity
  • No background metabolism for clear results

Single Overexpressed Human CYP450 Isozyme

CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over Human Liver Microsomes (HLMs) in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s or other classes of drug metabolizing enzymes.

High Activity Results in Increased Sensitivity

Compound rankings based on metabolism and inhibition profiles observed with CYP450 BACULOSOMES™ Plus Reagents are very similar to those seen with HLMs for most compounds tested. However, CYP450 BACULOSOMES™ Plus Reagents activity and metabolic rates with most substrates is significantly higher than those seen with HLMs. This results in a broad dynamic range and high sensitivity in assays utilizing CYP450 BACULOSOMES™ Plus Reagents. The high level of activity and reproducibility of the enzyme component observed in the reaction with most probe substrates makes CYP450 BACULOSOMES™ Plus Reagents well suited to high-throughput screening formats.

High Signal to Noise Ratio

No background metabolism by endogenous insect CYP450s has been detected with any of the Vivid™ probe substrates tested so far.

Applications

  • Isozyme identification
  • DMPK analysis
  • Isozyme-specific metabolism
  • Inhibition screening
TRUSTED_SUSTAINABILITY

Specifications

Description CYP2J2 BACULOSOMES™ Plus Reagent, 0.5nmol, Liquid, 3.3mg/mL Concentration, Cytochrome P450 Druggable Target, Microsomes Sub-Cellular Localization, Cytochrome P450 Druggable Target, Baculovirus Expression, Human Species, Used for Cell Viability, Proliferation and Function, Pharma and Biopharma
Concentration 3.3mg/mL
For Use With (Equipment) Microplate Reader
Product Line BACULOSOMES
Target Cytochrome P450
Formulation Liquid
For Use With (Application) Cell viability, proliferation and function, hepatocytes and hepatic models, pharma and biopharma, protein assays and analysis, target-based ADME/Tox assays
Sample Volume 0.5nmol
Content And Storage 1 tube, store at -80°C.
Product Type Bacterial Microsome
Quantity 0.5 nmol
Expression System Baculovirus
Protein Subtype Cytochrome P450 Enzymes
Shipping Condition Dry Ice
Sub Cellular Localization Microsomes
Research Category Toxicology & Drug Metabolism, Drug Development
Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.