missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP2F1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576223
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
CYP2F1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to dehydrogenate 3-methylindole, an endogenous toxin derived from the fermentation of tryptophan, as well as xenobiotic substrates such as naphthalene and ethoxycoumarin.
Specifications
| CYP2F1 | |
| Polyclonal | |
| Unconjugated | |
| CYP2F1 | |
| C2F1; CYP2F; Cyp2f1; Cyp2f2; Cyp2f4; CYP4502F4; CYPIIF1; CYPIIF2; Cytochrome P450 2F1; cytochrome P450 2F2; cytochrome P450 family 2 subfamily F member 1; cytochrome P450, family 2, subfamily F, polypeptide 1; cytochrome P450, family 2, subfamily f, polypeptide 2; cytochrome P450, family 2, subfamily f, polypeptide 4; cytochrome P450, subfamily 2F, polypeptide 1; cytochrome P450, subfamily IIF, polypeptide 1; cytochrome P450-NAH-2; flavoprotein-linked monooxygenase; microsomal monooxygenase; naphthalene dehydrogenase; Naphthalene hydroxylase; P450-NAH-2; xenobiotic monooxygenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1572, 54246 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35293, P24903 | |
| CYP2F1, Cyp2f4 | |
| Recombinant protein of human CYP2F1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction