missing translation for 'onlineSavingsMsg'
Learn More

CYP2F1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18643222 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18643222 0.02 mL 0.02mL
18655912 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18643222 Supplier Novus Biologicals Supplier No. NBP2928100.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYP2F1 Polyclonal antibody specifically detects CYP2F1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2F1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias C2F1, CYP2F, CYPIIF1, cytochrome P450 2F1, cytochrome P450, family 2, subfamily F, polypeptide 1, cytochrome P450, subfamily IIF, polypeptide 1, EC 1.14.14.1, flavoprotein-linked monooxygenase, MGC126121, microsomal monooxygenase, xenobiotic monooxygenase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-290 of human CYP2F1 (NP_000765.2). DYPAFFNFTKGNGIAFSSGDRWKVLRQFSIQILRNFGMGKRSIEERILEEGSFLLAELRKTEGEPFDPTFVLSRSVSNIICSVLFGSRFDYDDERLLTIIRLINDNFQIMSSPWGELYDIFPSLLDWVPGPHQRIFQNFKCLRDLIAHSVHDHQASLDPRSPRDFIQCFLTKMAEEKEDPLSHFHMDTLLM
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1572
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.