missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP2C9 BACULOSOMES™ Plus Reagent, rHuman

Product Code. 10013092
Change view
Click to view available options
Quantity:
0.5 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10013092 0.5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10013092 Supplier Invitrogen™ Supplier No. P2378

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CYP2C9 BACULOSOMES™ Plus Reagent, rHuman

Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase. For this particular isozyme (CYP2C9), human cytochrome b5 is also included.
  • Single human P450 isozyme for detailed drug metabolism studies
  • High specific activity increases dynamic range and sensitivity
  • No background metabolism for clear results

    Single Overexpressed Human CYP450 Isozyme
    CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over Human Liver Microsomes (HLMs) in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s or other classes of drug metabolizing enzymes.

    High Activity Results in Increased Sensitivity
    Compound rankings based on metabolism and inhibition profiles observed with CYP450 BACULOSOMES™ Plus Reagents are very similar to those seen with HLMs for most compounds tested. However, CYP450 BACULOSOMES™ Plus Reagents activity and metabolic rates with most substrates is significantly higher than those seen with HLMs. This results in a broad dynamic range and high sensitivity in assays utilizing CYP450 BACULOSOMES™ Plus Reagents. The high level of activity and reproducibility of the enzyme component observed in the reaction with most probe substrates makes CYP450 BACULOSOMES™ Plus Reagents well suited to high-throughput screening formats.

    High Signal to Noise Ratio
    No background metabolism by endogenous insect CYP450s has been detected with any of the Vivid™ probe substrates tested so far.

    Applications: isozyme identification, DMPK analysis, isozyme-specific metabolism, and inhibition screening

    For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Host Species Human
    Content And Storage -80°C
    Shipping Condition Dry Ice
    Quantity 0.5 nmol
    Product Type Reagent
    Research Category Toxicology and Drug Metabolism
    Product Line BACULOSOMES
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.