missing translation for 'onlineSavingsMsg'
Learn More

CYP2A7 Antibody, Novus Biologicals™

Product Code. 18364508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364508 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364508 Supplier Novus Biologicals Supplier No. H00001549B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CYP2A7 Polyclonal antibody specifically detects CYP2A7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2A7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_000755
Gene Alias CPA7, CPAD, CYP2A, CYPIIA7, cytochrome P450 2A7, Cytochrome P450 IIA4, cytochrome P450, family 2, subfamily A, polypeptide 7, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 7, EC 1.14.14.1, P450-IIA4
Host Species Mouse
Immunogen CYP2A7 (NP_000755, 1 a.a. - 494 a.a.) full-length human protein. MLASGLLLVALLACLTVMVLMSVWQQRKSRGKLPPGPTPLPFIGNYLQLNTEHICDSIMKFSECYGPVFTIHLGPRRVVVLCGHDAVREALVDQAEEFSGRGEQATFDWVFKGYGVAFSNGERAKQLLRFAIATLRDFGVGKRGIEERIQEESGFLIEAIRSTHGANIDPTFFLSRTVSNVISSIVFGDRFDYEDKEFLSLLSMMLGIFQFTSTSTGQLYEMFSSVMKHLPGPQQQAFKLLQGLEDFIAKKVEHNQRTLDPNSPQDFIDSFLIHMQEEEKNPNTEFYLKNLMMSTLNLFIAGTETVSTTLRYGFLLLMKHPEVEAKVHEEIDRVIGKNRQPKFEDRTKMPYMEAVIHEIQRFGDVIPMSLARRVKKDTKFRDFFLPKGTEVFPMLGSVLRDPSFFSNPQDFNPQHFLDDKGQFKKSDAFVPFSIGKRNCFGEGLARMELFLFFTTVMQNFRLKSSQSPKDIDVSPKHVVFATIPRNYTMSFLPR
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1549
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.