missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2A6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21721-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene, CYP2A6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to hydroxylate coumarin, and also metabolizes nicotine, aflatoxin B1, nitrosamines, and some pharmaceuticals. Individuals with certain allelic variants are said to have a poor metabolizer phenotype, meaning they do not efficiently metabolize coumarin or nicotine. This gene is part of a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q. The gene was formerly referred to as CYP2A3; however, it has been renamed CYP2A6.Specifications
| CYP2A6 | |
| Polyclonal | |
| Unconjugated | |
| CYP2A6 | |
| Coumarin 7 hydroxylase, CPA6, CYP2A, CYP2A3, CYP2A6, CYPIIA6, Cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), P450C2A, P450PB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110690, 1548, 29594 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11509 | |
| CYP2A6 | |
| CYP2A6 Fusion Protein Ag16377 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction