missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2A6, Mouse anti-Human, Clone: 3B11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001548-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LGSVLRDPSFFSNPQDFNPQHFLNEKGQFKKSDAFVPFSIGKRNCFGEGLARMELFLFFTTVMQNFRLKSSQSPKDIDVSPKHVGFATIPRNYTMSFLPRSpecifications
| CYP2A6 | |
| Monoclonal | |
| Unconjugated | |
| cytochrome P450, family 2, subfamily A, polypeptide 6 | |
| CPA6/CYP2A/CYP2A3/P450C2A/P450PB | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B11 | |
| In 1x PBS, pH 7.4 | |
| NG_000008 | |
| CYP2A6 | |
| CYP2A6 (AAH28215, 395 a.a. ∼ 494 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1548 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction