missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP27C1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567008
This item is not returnable.
View return policy
Description
Immunogen sequence: FPVLPGNGRVT QEDLVIGGYL IPKGTQLALC HYATSYQDEN FPRAKEFRPE RWLRKGDLDR VDNFGSIPFG HGVRS Highest antigen sequence identity to the following orthologs - mouse 48%, rat 48%.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.
Specifications
| CYP27C1 | |
| Polyclonal | |
| Unconjugated | |
| CYP27C1 | |
| All-trans retinol 3,4-desaturase; CYP27C1; Cytochrome P450 27C1; cytochrome P450 family 27 subfamily C member 1; cytochrome P450, family 27, subfamily C, polypeptide 1; cytochrome P450, family 27, subfamily C, polypeptide 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 339761 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4G0S4 | |
| CYP27C1 | |
| Recombinant Human CYP27C1. Recombinant protein control fragment (Product #RP-107648). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction