missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CYP27A1 Monoclonal specifically detects CYP27A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
| Antigen | CYP27A1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | V29-P4B8*B2 |
| Conjugate | DyLight 755 |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | 12-alpha-triol 27-hydroxylase, 7-alpha, cholestanetriol 26-monooxygenase, CP27,5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase, CTX5-beta-cholestane-3-alpha, CYP275-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase, Cytochrome P450 27, cytochrome P450, family 27, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinousxanthomatosis), polypeptide 1, Cytochrome P-450C27/25, EC 1.14.13.15, sterol 26-hydroxylase, mitochondrial, Sterol 27-hydroxylase, Vitamin D(3) 25-hydroxylase |
| Gene Symbols | CYP27A1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?