missing translation for 'onlineSavingsMsg'
Learn More

CYP27A1 Antibody (V29-P4B8*B2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-76660-0.025ml

 View more versions of this product

Product Code. 18604829

  • £150.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CYP27A1 Monoclonal specifically detects CYP27A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CYP27A1
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA, Immunohistochemistry 1:10 - 1:500
12-alpha-triol 27-hydroxylase, 7-alpha, cholestanetriol 26-monooxygenase, CP27,5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase, CTX5-beta-cholestane-3-alpha, CYP275-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase, Cytochrome P450 27, cytochrome P450, family 27, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinousxanthomatosis), polypeptide 1, Cytochrome P-450C27/25, EC 1.14.13.15, sterol 26-hydroxylase, mitochondrial, Sterol 27-hydroxylase, Vitamin D(3) 25-hydroxylase
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
V29-P4B8*B2
Unconjugated
PBS with 0.02% sodium azide
CYP27A1
Ovalbumin-conjugated synthetic peptide KVVLAPETGEL (C-terminal sequence)
0.025 mL
Stem Cell Markers
1593
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.