missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CYP26C1 Monoclonal antibody specifically detects CYP26C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | CYP26C1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | T6P1C7*E7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Formulation | PBS |
| Gene Alias | cytochrome P450 26C1, cytochrome P450, family 26, subfamily C, polypeptide 1, EC 1.14, FLJ45301 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?