missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP26C1 Antibody (T6P1C7*E7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £302.00
Specifications
| Antigen | CYP26C1 |
|---|---|
| Clone | T6P1C7*E7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687227
|
Novus Biologicals
NBP2-50196-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675286
|
Novus Biologicals
NBP2-50196 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP26C1 Monoclonal antibody specifically detects CYP26C1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| CYP26C1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| metabolism, Signal Transduction | |
| PBS | |
| 340665 | |
| IgG1 κ | |
| Protein A purified |
| T6P1C7*E7 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cytochrome P450 26C1, cytochrome P450, family 26, subfamily C, polypeptide 1, EC 1.14, FLJ45301 | |
| Ovalbumin-conjugated synthetic peptide RWELATPAFP (C-terminal sequence) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title