missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP26B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21555-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and the synthesis of cholesterol, steroids and other lipids. The enzyme encoded by this gene is involved in the specific inactivation of all-trans-retinoic acid to hydroxylated forms, such as 4-oxo-, 4-OH-, and 18-OH-all-trans-retinoic acid.Specifications
| CYP26B1 | |
| Polyclonal | |
| Unconjugated | |
| CYP26B1 | |
| CYP26A2, CYP26B1, Cytochrome P450 26A2, Cytochrome P450 26B1, Cytochrome P450RAI 2, P450RAI 2, P450RAI2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 232174, 312495, 56603 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| G3V7X8, Q811W2, Q9NR63 | |
| Cyp26b1 | |
| CYP26B1 Fusion Protein Ag15368 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction