missing translation for 'onlineSavingsMsg'
Learn More

CYP24A1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16871185 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16871185 20 μL 20µL
16861185 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16871185 Supplier Proteintech Supplier No. 215821AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CYP24A1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CYP24A1. This antibody recognizes human, mouse, rat antigen. The CYP24A1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.

Specifications

Antigen CYP24A1
Applications Western Blot, Immunoprecipitation, Immunohistochemistry
Classification Polyclonal
Concentration 400 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1591
Target Species Human, Rat, Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.