missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP24A1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519406
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This mitochondrial protein initiates the degradation of 1,25-dihydroxyvitamin D3, the physiologically active form of vitamin D3, by hydroxylation of the side chain. In regulating the level of vitamin D3, this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| CYP24A1 | |
| Polyclonal | |
| Unconjugated | |
| Cyp24a1 | |
| 1,25-@dihydroxyvitamin D3 24-hydroxylase; 1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial; 24-OHase; 24R-Ohase; 25-hydroxyvitamin D-24-hydroxylase; 25-hydroxyvitamin D3 24-hydroxylase; 25-hydroxyvitaminD324-hydroxylase; CP24; CYP24; Cyp-24; CYP24A1; Cytochrom P450 (25-hydroxyvitamin D24-hydroxylase); cytochrome P450 24A1; cytochrome P450 family 24 subfamily A member 1; cytochrome P450, 24; cytochrome P450, 24a1; cytochrome P450, family 24, subfamily a, polypeptide 1; cytochrome P450, subfamily 24; cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase); cytochrome P450-CC24; exo-mitochondrial protein; HCAI; HCINF1; MGC126273; MGC126274; P450-CC24; vitamin D 24-hydroxylase; vitamin D(3) 24-hydroxylase; VITD12 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1591, 25279 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q07973, Q09128 | |
| Cyp24a1 | |
| Synthetic peptide sequence (RKYDIQATDN) corresponding to the internal amino acids of CYP24A1 (aa 481-490). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction