Learn More
CYP24A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521704
Description
PA5-21704 targets CYP24A1 in WB and IHC-P applications and shows reactivity with Human samples. The PA5-21704 immunogen is a recombit fragment corresponding to a region within amino acids 241 and 514 of CYP24A1 (Uniprot IDNo.Q07973). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
| CYP24A1 | |
| Polyclonal | |
| Unconjugated | |
| Q07973, Q64441 | |
| CYP24A1 | |
| Recombit fragment corresponding to a region within amino acids 241 and 514 of CYP24A1 (Uniprot IDNo.Q07973) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| CYP24A1 | |
| CP24, CYP24, HCAI, P450-CC24, 1,25-@dihydroxyvitamin D3 24-hydroxylase, 1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial, 24-OHase, cytochrome P450 24A1, cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase), cytochrome P450-CC24, exo-mitochondrial protein, vitamin D 24-hydroxylase, vitamin D(3) 24-hydroxylase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13081, 1591 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.