missing translation for 'onlineSavingsMsg'
Learn More

CYP24A1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18350516 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350516 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350516 Supplier Novus Biologicals Supplier No. NBP305981JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYP24A1 Polyclonal specifically detects CYP24A1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Spécification

Antigen CYP24A1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial, CP241, CYP241,25-alphadihydroxyvitamin D3 24-hydroxylase, Cytochrome P450 24A1, cytochrome P450, family 24, subfamily A, polypeptide 1,24-OHase, cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase), Cytochrome P450-CC24, EC 1.14.13.n4, exo-mitochondrial protein, MGC126273, MGC126274, P450-CC24, vitamin D 24-hydroxylase, Vitamin D(3) 24-hydroxylase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CYP24A1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1591
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.