missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP20A1 Antibody (Z27-P3H7*E3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £315.00
Specifications
| Antigen | CYP20A1 |
|---|---|
| Clone | Z27-P3H7*E3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639656
|
Novus Biologicals
NBP2-50192-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638997
|
Novus Biologicals
NBP2-50192 |
0.1 mL |
£315.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP20A1 Monoclonal antibody specifically detects CYP20A1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP20A1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| PBS | |
| 57404 | |
| IgG | |
| Protein A purified |
| Z27-P3H7*E3 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CYP-M, cytochrome P450 20A1, cytochrome P450 monooxygenase, cytochrome P450, family 20, subfamily A, polypeptide 1, EC 1.14, MGC22229 | |
| Ovalbumin-conjugated synthetic peptide AWITVSKRY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title