missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP1B1 Recombinant Rabbit Monoclonal Antibody (K01_1Q09)

Product Code. 30829398
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829398 100 μL
30825038 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829398 Supplier Invitrogen™ Supplier No. MA563709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: NHDPLKWPNP ENFDPARFLD KDGLINKDLT SRVMIFSVGK RRCIGEELSK MQLFLFISIL AHQCDFRANP NEPAKMNFSY GLTIKPKSFK VNVTLRESME LLDSAVQNLQ AKETCQ Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%, Pig - 84%, Cynomolgus monkey - 94%.

CYP1B1 is a 543 amino acid intracellular protein localized to the endoplasmic reticulum. It belongs to the cytochrome P450 super family of enzymes which are monooxygenases and it catalyzes many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme metabolizes procarcinogens such as polycyclic aromatic hydrocarbons and 17beta-estradiol and is also involved in the metabolism of substances active in the eye growth and differentiation and hence plays a putative role in ocular development. Mutated proteins are known to cause primary congenital glaucoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP1B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q09
Concentration 1.30 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Cyp1b1
Gene Accession No. Q16678, Q64678
Gene Alias aryl hydrocarbon hydroxylase; benz[a]anthracene inducible; CP1B; CYP1B; CYP1B1; Cyp1-b1; CYPIB1; cytochrome P450 1B; cytochrome P450 1B1; cytochrome P450 family 1 subfamily B member 1; cytochrome P450, 1b1, benz[a]anthracene inducible; cytochrome P450, family 1, subfamily B, polypeptide 1; cytochrome P450, subfamily 1B, polypeptide 1; cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, primary infantile); cytochrome P4501B; cytochrome P450CMEF; Cytochrome P450EF; cytochrome P450RAP; flavoprotein-linked monooxygenase; GLC3A; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P4501B1; P450RAP; xenobiotic monooxygenase; zgc:136223
Gene Symbols Cyp1b1
Host Species Rabbit
Immunogen Recombinant protein of human CYP1B1 (aa 428-543).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1545, 25426
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.