missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Mouse anti-Human, Unconjugated, Clone: 1G8D4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CYP1B1 |
|---|---|
| Clone | 1G8D4 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889415
|
Proteintech
67033-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879415
|
Proteintech
67033-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CYP1B1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP1B1. This antibody recognizes human, mouse, rat, pig antigen. The CYP1B1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CYP1B1 | |
| 500 μg/mL | |
| Monoclonal | |
| Mouse | |
| 1545 | |
| Primary |
| 1G8D4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Mouse, Rat, Pig | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title