missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Antibody (S02-5A3), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £516.00
Specifications
| Antigen | CYP1B1 |
|---|---|
| Clone | S02-5A3 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot 1/2000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685164
|
Novus Biologicals
NBP3-19616-100ul |
100 μg |
£516.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663975
|
Novus Biologicals
NBP3-19616-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP1B1 Monoclonal antibody specifically detects CYP1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CYP1B1 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 1545 | |
| IgG | |
| Affinity purified |
| S02-5A3 | |
| Western Blot 1/2000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase | |
| Recombinant protein of human CYP1B1 (Uniprot # Q16678) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title